Potassium Bromide Crystal Optic Disc
Catalog # BT123425-1EACH
Supplier: BeanTown Chemical
Specifications
- Description:Potassium bromide crystal (MFCD00011358), CAS No. 7758-02-3
- Dimensions:25×4 mm
- Cat. No.:BT123425-1EACH
- Supplier no.:123425-1EACH
Specifications
About this item
Form: Crystalline
Signal word: Danger
GHS symbol: GHS06
EC Number: 231-830-3
Melting point/Melting range: 734 °C (1353.2 °F)
Boiling point/Boiling range: 1.435 °C (34.635 °F)
Density at 20 °C (68 °F): 2.75 g/cm³ (22.94875 lbs/gal)
Solids content: 100.0%
Delivery: Non-hazardous for shipping.