Bromide, Ion Chromatography Standard Solution, Br⁻ 1,000 µg/ml
Supplier: BeanTown Chemical
Certificates
About this item
Solids content: 0.2%
Density at 20 °C (68 °F): 1 g/cm³ (8.345 lbs./gal)
Solubility in/Miscibility with water: Fully miscible
Vapor pressure at 20 °C (68 °F): 23 hPa (17.3 mm Hg)
- Solvent content:
- Water: 99.9 %
- VOC content: 0.00%
- 0.0 g/L /0.00 lbs./gal
Delivery information: Non-hazardous for shipping.
Specifications
- MDL No.:MFCD00134172
- Physical Form:Liquid
Specifications
Frequently Bought Together