Go Wireless® Heart Rate
Catalog # 470165-082
Supplier: Vernier Software & Technology
Specifications
- Interface:
- Description:Go Wireless® heart rate
- Environmentally Preferable:
- Cat. No.:470165-082
- Supplier no.:GW-HR
Specifications
About this item
The Vernier Go Wireless® Heart Rate is ideal for continuously monitoring heart rate before, during, and after exercise or while a person is stationary.
- Hands-free monitoring
- For heart-rate monitoring
- Wireless transmission
- Simple and easy to use
- Displays live heart rate readouts before data collection
- Hand grips do not require electrodes or clips
- Easy to clean and share between students
Data is wirelessly transmitted to iPad®, LabQuest 2, or Android™ devices.
Delivery: Includes Heart rate hand grips, polar transmittor module.