Exercise Heart Rate Monitor
Catalog # 470165-186
Supplier: Vernier Software & Technology
Specifications
- Description:Exercise heart rate monitor
- Cat. No.:470165-186
- Supplier no.:GW-EHR
Specifications
About this item
The Vernier Exercise Heart Rate Monitor is an excellent hands-free option for continuously monitoring heart rate before, during, and after exercise or while a person is stationary.
- Hands-free monitoring
- For heart-rate monitoring
- Wireless transmission
Data is wirelessly transmitted to iPad®, LabQuest 2, or Android™ devices. Chest strap is ideal for running, cycling, and other physical activities.
Delivery: Includes Exercise heart rate strap, polar transmittor module.