DETA NONOate (NOC-18)
Supplier: Avantor
Certificates
About this item
CAS Number:146724-94-9
:
Formula:C₄H₁₃N₅O₂
MDL Number:MFCD00278808
:163.181 g/mol
Specification Test Results
- CAS:146724-94-9
- Purity:≥98%
- Form:White solid
Specification Test Results
Frequently Bought Together