Thionitrobenzoic acid (TNB) 95%
Supplier: Avantor
Certificates
About this item
CAS Number:15139-21-6
:
Formula:C₇H₅NO₄S
MDL Number:MFCD14641982
:199.18 g/mol
Storage Temperature:Freezer
Specification Test Results
- CAS:15139-21-6
- Purity:≥98% by HPLC
- Form:Yellow solid
Specification Test Results
Frequently Bought Together