1,5-Isoquinolinediol
Supplier: Avantor
Certificates
About this item
CAS Number:5154-02-9
:
Formula:C₉H₇NO₂
MDL Number:MFCD00006900
:161.16 g/mol
Specification Test Results
- CAS:1188540
- Purity:≥98% by TLC
- Form:Off-white solid
Specification Test Results
Frequently Bought Together