11-(1-Piperazinyl)dibenzo[b,f][1,4]thiazepine dihydrochloride
Supplier: Avantor
Certificates
About this item
CAS Number:111974-74-4
:
Formula:C₁₇H₁₉Cl₂N₃S
MDL Number:MFCD08703302
:368.3239 g/mol
Specification Test Results
- CAS:111974-74-4
- Purity:≥98% TLC
- Form:Off-white solid
Specification Test Results
Frequently Bought Together