Sulfo-SMCC (3-Sulfo-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt)
Supplier: Avantor
Certificates
About this item
CAS Number:92921-24-9
:
Formula:C₁₆H₁₈N₂O₉S
MDL Number:MFCD00054978
:436.37 g/mol
Specification Test Results
- CAS:92921-24-9
- Purity:≥90% by NMR
- Form:White to off-white solid
Specification Test Results
Frequently Bought Together