Cathepsin G inhibitor I
Catalog # 10188-642
Supplier: Avantor
CAS Number:
Specifications
- Size:1 mg
- Cat. No.:10188-642
- Supplier no.:1982-1
Specifications
Specification Test Results
- CAS:429676-93-7
- Purity:≥98% by HPLC
- Form:White solid
Specification Test Results
About this item
:
:620.60 g/mol
CAS Number:429676-93-7
Formula:C₃₆H₃₃N₂O₆P
MDL Number:MFCD06412001