Elastase Inhibitor
Catalog # 10188-514
Supplier: Avantor
CAS Number:
Specifications
- CAS No.:111682-13-4
- Size:5 mg
- Protein/Peptide Name:Elastase Inhibitor
- Cat. No.:10188-514
- Supplier no.:1922-5
Specifications
About this item
- White solid
- Purity: ≥98% by TLC