ContactUsLinkComponent
Neulog Drop Counter
Neulog Drop Counter
Catalog # 470122-798
Supplier:  Wards
undefined
Neulog Drop Counter
Catalog # 470122-798
Supplier:  Wards
Supplier Number:  NUL223
Specifications
  • Resolution:
    1
    ADC: 10 bit
  • Description:
    Neulog drop counter
  • Measurement Time:
    1 s - 31 days
  • Cat. No.:
    470122-798
  • Range:
    0 - 6500 drops
  • Supplier no.:
    NUL223
Specifications
About this item
Take the human error out of counting drops and bubbles during experiments and demos.
  • Max. sample rate 100 S/sec.
This sensor enables to count automatically drops and bubbles. It is perfect for titrations. Combined with the pH sensor, it gives a very nice titration curve.
Ordering information: For each probe you will need one or more of the following items: Battery Module, Viewer module (not needed if connecting to a computer to display data), USB module (not needed if only using as a stand alone probe or you have the WIFI module), WIFI module (Not needed if connecting to a computer through the USB module). Recommended is to have a battery module and a USB module.
Caution: For educational use only.