Order Entry
United States
ContactUsLinkComponent
Precision Air Table
Precision Air Table
Catalog # 470157-648
Supplier:  Wards
undefined
Precision Air Table
Catalog # 470157-648
Supplier:  Wards
Supplier Number:  AIRTBL

Specifications

  • Description:
    Precision Air Table
  • Cat. No.:
    470157-648
  • Supplier no.:
    AIRTBL

Specifications

About this item

The air table guides the study of a multitude of concepts.

  • Topics Include Elastic and Inelastic Collisions, Linear and Angular Momentum, and More
  • Air Table Length: 700 mm
  • Width: 500 mm
  • Adjustable Feet

These concepts include elastic and inelastic collisions, linear and angular momentum, conservation of momentum, moment of inertia, centripetal force, and simple and complex harmonic motion.

The air table is 500 x 700 mm with adjustable feet. It requires an air blower.