Temp-Tec NL-56, Knitted Neoprene Gloves, MAPA® Professional
Supplier: MAPA
Certificates
About this item
Gloves are chemical-resistant and provide thermal insulation.
- Insulated neoprene gloves
- Positive non-slip roughed grip
- Two-color construction for a visual indicator of glove wear-through
Gloves offer reliable hand protection for chemical processing, metal finishing, road works, polymer extrusion, Fish filleting and mould handling in ceramic industry.
Certifications: In compliance with 21 CFR 170-199.
Specifications
- Catalog No:
- 13450-196
- 32920-012
- 32920-017
- Ambidextrous/Hand-specific:
- Hand-Specific
- Hand-Specific
- Hand-Specific