Order Entry
United States
Orders LinkContactUsLinkComponent
 

iMark™ Microplate Absorbance Reader, Bio-Rad

Marketsource
Supplier:   Bio-Rad
iMark™ Microplate Absorbance Reader, Bio-Rad
Supplier:  Bio-Rad
Marketsource
Certificates

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

The iMark™ Microplate Absorbance Reader, with a wavelength range of 400–750 nm, is an economical solution for a wide range of applications, including immunoassays with colorimetric substrates, such as ELISA and protein assays

  • Fast microplate reading speeds
  • Convenient built-in shaker
  • Internal wide-format thermal graphical printer
  • Easy-to-use, stand-alone operation
  • Extensive onboard assay configuration
  • Onboard software — includes programmable kinetic reading, comprehensive curve-fit selections
  • Security login that allows locking and unlocking of key protocol settings by the administrator
  • Checkmark™ Reader Performance Verification Kit has preprogrammed protocols to validate the IQ/OQ/PQ in compliance with GLP
  • Ordering information: The iMark™ Microplate Absorbance Reader includes 415, 450, 490, 595, 655, and 750 nm filters, 8-filter capacity filter wheel, built-in plate shaker, onboard software and thermal printer, one roll printer paper, USB, and power cables. Microplate Manager® 6 Software is included with 1681135. A large selection of additional absorbance filters, mostly at 5 nm intervals from 400 to 700 nm, are available (see Accessories tab). The Model 1575 Immunowash™ Microplate Washer includes microplate washer, bottle, tubing, 8-port manifold, and aerosol protection cover.

The instrument can be used as a stand-alone reader using the 64 built-in assay protocols, data storage, analysis, and reports. The Microplate Manager® Software (PC or Mac) can be used to control the reader and format readings, plus more extensive data analysis.

Specifications for Product Family

Specifications

  • Catalog No:
  • 100069-868
  • 100447-556
  • 100447-558
  • 100447-570
  • 100447-572
  • Wavelength:
  • 400–750 nm
  • 400–750 nm
  • 400–750 nm
  • 400–750 nm
  • 400–750 nm
  • Accuracy:
  • ≤1.0% or 0.010 from 0.000–3.000 OD at 490 nm
  • ≤1.0% or 0.010 from 0.000–3.000 OD at 490 nm
  • ≤1.0% or 0.010 from 0.000–3.000 OD at 490 nm
  • ≤1.0% or 0.010 from 0.000–3.000 OD at 490 nm
  • ≤1.0% or 0.010 from 0.000–3.000 OD at 490 nm
  • Output:
  • Onboard graphical thermal printer and USB2 interface with PC or Mac data stations
  • Onboard graphical thermal printer and USB2 interface with PC or Mac data stations
  • Onboard graphical thermal printer and USB2 interface with PC or Mac data stations
  • Onboard graphical thermal printer and USB2 interface with PC or Mac data stations
  • Onboard graphical thermal printer and USB2 interface with PC or Mac data stations
  • Data Storage:
  • Calender/clock functions; 64 assay protocols
  • Calender/clock functions; 64 assay protocols
  • Calender/clock functions; 64 assay protocols
  • Calender/clock functions; 64 assay protocols
  • Calender/clock functions; 64 assay protocols
  • Duration:
  • 0 - 999 s
  • 0 - 999 s
  • 0 - 999 s
  • 0 - 999 s
  • 0 - 999 s
  • Filters:
  • 415, 450, 490, 595, 655, and 750 nm
  • 415, 450, 490, 595, 655, and 750 nm
  • 415, 450, 490, 595, 655, and 750 nm
  • 415, 450, 490, 595, 655, and 750 nm
  • 415, 450, 490, 595, 655, and 750 nm
  • Languages:
  • 4 languages (English, Chinese, Japanese, Russian); LCD indication supported; printout report supported
  • 4 languages (English, Chinese, Japanese, Russian); LCD indication supported; printout report supported
  • 4 languages (English, Chinese, Japanese, Russian); LCD indication supported; printout report supported
  • 4 languages (English, Chinese, Japanese, Russian); LCD indication supported; printout report supported
  • 4 languages (English, Chinese, Japanese, Russian); LCD indication supported; printout report supported
  • Linearity:
  • ≤1.0% from 0.0–2.0 OD; ≤2.0% from 0.0–3.0 OD
  • ≤1.0% from 0.0–2.0 OD; ≤2.0% from 0.0–3.0 OD
  • ≤1.0% from 0.0–2.0 OD; ≤2.0% from 0.0–3.0 OD
  • ≤1.0% from 0.0–2.0 OD; ≤2.0% from 0.0–3.0 OD
  • ≤1.0% from 0.0–2.0 OD; ≤2.0% from 0.0–3.0 OD
  • Precision:
  • 1.0% or 0.005 OD from 0.0–2.0 OD; 1.5% from 2.0–3.0 OD
  • 1.0% or 0.005 OD from 0.0–2.0 OD; 1.5% from 2.0–3.0 OD
  • 1.0% or 0.005 OD from 0.0–2.0 OD; 1.5% from 2.0–3.0 OD
  • 1.0% or 0.005 OD from 0.0–2.0 OD; 1.5% from 2.0–3.0 OD
  • 1.0% or 0.005 OD from 0.0–2.0 OD; 1.5% from 2.0–3.0 OD
  • Printer:
  • Built-in, thermal
  • Built-in, thermal
  • Built-in, thermal
  • Built-in, thermal
  • Built-in, thermal
  • Range:
  • 0.0–3.5 OD
  • 0.0–3.5 OD
  • 0.0–3.5 OD
  • 0.0–3.5 OD
  • 0.0–3.5 OD
  • Resolution:
  • 0.001 OD
  • 0.001 OD
  • 0.001 OD
  • 0.001 OD
  • 0.001 OD
  • Shaking Speed:
  • Low, mid, high
  • Low, mid, high
  • Low, mid, high
  • Low, mid, high
  • Low, mid, high
  • Time:
  • 6 sec at single wavelength, 10 sec at dual wavelengths
  • 6 sec at single wavelength, 10 sec at dual wavelengths
  • 6 sec at single wavelength, 10 sec at dual wavelengths
  • 6 sec at single wavelength, 10 sec at dual wavelengths
  • 6 sec at single wavelength, 10 sec at dual wavelengths
  • Voltage:
  • 100/240 V
  • 100/240 V
  • 100/240 V
  • 100/240 V
  • 100/240 V
  • W×D×H:
  • 34.6×37.7×16.4 cm (13.6×14.8×6.5")
  • 34.6×37.7×16.4 cm (13.6×14.8×6.5")
  • 34.6×37.7×16.4 cm (13.6×14.8×6.5")
  • 34.6×37.7×16.4 cm (13.6×14.8×6.5")
  • 34.6×37.7×16.4 cm (13.6×14.8×6.5")
  • Weight:
  • 5.5 kg (12 lbs.)
  • 5.5 kg (12 lbs.)
  • 5.5 kg (12 lbs.)
  • 5.5 kg (12 lbs.)
  • 5.5 kg (12 lbs.)

Specifications

Product Family Options

Product Information

  • Availability
    Price
  • 100069-868
    Model 1575 Immunowash™ Microplate Washer, Programmable up to 75 Sequences
    Model 1575 Immunowash™ Microplate Washer, Programmable up to 75 Sequences
    Log in for availability
    Log in for availability
    Each
    Each - $16,278.46
    $16,278.46
    Each
       $16,278.46

    Supplier Number: 1707009
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    100069-868
    Wavelength
    400–750 nm
    Accuracy
    ≤1.0% or 0.010 from 0.000–3.000 OD at 490 nm
    Output
    Onboard graphical thermal printer and USB2 interface with PC or Mac data stations
    Data Storage
    Calender/clock functions; 64 assay protocols
    Duration
    0 - 999 s
    Filters
    415, 450, 490, 595, 655, and 750 nm
    Languages
    4 languages (English, Chinese, Japanese, Russian); LCD indication supported; printout report supported
    Linearity
    ≤1.0% from 0.0–2.0 OD; ≤2.0% from 0.0–3.0 OD
    Precision
    1.0% or 0.005 OD from 0.0–2.0 OD; 1.5% from 2.0–3.0 OD
    Printer
    Built-in, thermal
    Range
    0.0–3.5 OD
    Resolution
    0.001 OD
    Shaking Speed
    Low, mid, high
    Time
    6 sec at single wavelength, 10 sec at dual wavelengths
    Voltage
    100/240 V
    W×D×H
    34.6×37.7×16.4 cm (13.6×14.8×6.5")
    Weight
    5.5 kg (12 lbs.)
    Marketsource
  • 100447-556
    iMark™ Microplate Absorbance Reader
    iMark™ Microplate Absorbance Reader
    Discontinued
    Supplier Number: -
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    100447-556
    Wavelength
    400–750 nm
    Accuracy
    ≤1.0% or 0.010 from 0.000–3.000 OD at 490 nm
    Output
    Onboard graphical thermal printer and USB2 interface with PC or Mac data stations
    Data Storage
    Calender/clock functions; 64 assay protocols
    Duration
    0 - 999 s
    Filters
    415, 450, 490, 595, 655, and 750 nm
    Languages
    4 languages (English, Chinese, Japanese, Russian); LCD indication supported; printout report supported
    Linearity
    ≤1.0% from 0.0–2.0 OD; ≤2.0% from 0.0–3.0 OD
    Precision
    1.0% or 0.005 OD from 0.0–2.0 OD; 1.5% from 2.0–3.0 OD
    Printer
    Built-in, thermal
    Range
    0.0–3.5 OD
    Resolution
    0.001 OD
    Shaking Speed
    Low, mid, high
    Time
    6 sec at single wavelength, 10 sec at dual wavelengths
    Voltage
    100/240 V
    W×D×H
    34.6×37.7×16.4 cm (13.6×14.8×6.5")
    Weight
    5.5 kg (12 lbs.)
    Marketsource
  • 100447-558
    iMark™ Microplate Absorbance Reader with Microplate Manager® 6
    iMark™ Microplate Absorbance Reader with Microplate Manager® 6
    Discontinued
    Supplier Number: -
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    100447-558
    Wavelength
    400–750 nm
    Accuracy
    ≤1.0% or 0.010 from 0.000–3.000 OD at 490 nm
    Output
    Onboard graphical thermal printer and USB2 interface with PC or Mac data stations
    Data Storage
    Calender/clock functions; 64 assay protocols
    Duration
    0 - 999 s
    Filters
    415, 450, 490, 595, 655, and 750 nm
    Languages
    4 languages (English, Chinese, Japanese, Russian); LCD indication supported; printout report supported
    Linearity
    ≤1.0% from 0.0–2.0 OD; ≤2.0% from 0.0–3.0 OD
    Precision
    1.0% or 0.005 OD from 0.0–2.0 OD; 1.5% from 2.0–3.0 OD
    Printer
    Built-in, thermal
    Range
    0.0–3.5 OD
    Resolution
    0.001 OD
    Shaking Speed
    Low, mid, high
    Time
    6 sec at single wavelength, 10 sec at dual wavelengths
    Voltage
    100/240 V
    W×D×H
    34.6×37.7×16.4 cm (13.6×14.8×6.5")
    Weight
    5.5 kg (12 lbs.)
    Marketsource
  • 100447-570
    Checkmark™ Reader Performance Verification Kit (for GLP compliance)
    Checkmark™ Reader Performance Verification Kit (for GLP compliance)
    Log in for availability
    Log in for availability
    Each
    Each - $5,772.31
    $5,772.31
    Each
       $5,772.31

    Supplier Number: 1686940
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    100447-570
    Wavelength
    400–750 nm
    Accuracy
    ≤1.0% or 0.010 from 0.000–3.000 OD at 490 nm
    Output
    Onboard graphical thermal printer and USB2 interface with PC or Mac data stations
    Data Storage
    Calender/clock functions; 64 assay protocols
    Duration
    0 - 999 s
    Filters
    415, 450, 490, 595, 655, and 750 nm
    Languages
    4 languages (English, Chinese, Japanese, Russian); LCD indication supported; printout report supported
    Linearity
    ≤1.0% from 0.0–2.0 OD; ≤2.0% from 0.0–3.0 OD
    Precision
    1.0% or 0.005 OD from 0.0–2.0 OD; 1.5% from 2.0–3.0 OD
    Printer
    Built-in, thermal
    Range
    0.0–3.5 OD
    Resolution
    0.001 OD
    Shaking Speed
    Low, mid, high
    Time
    6 sec at single wavelength, 10 sec at dual wavelengths
    Voltage
    100/240 V
    W×D×H
    34.6×37.7×16.4 cm (13.6×14.8×6.5")
    Weight
    5.5 kg (12 lbs.)
    Marketsource
  • 100447-572
    Microplate Manager® 6, Version 6.3
    Microplate Manager® 6, Version 6.3
    Log in for availability
    Log in for availability
    Each
    Each - $4,727.69
    $4,727.69
    Each
       $4,727.69

    Supplier Number: 1689520
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    100447-572
    Wavelength
    400–750 nm
    Accuracy
    ≤1.0% or 0.010 from 0.000–3.000 OD at 490 nm
    Output
    Onboard graphical thermal printer and USB2 interface with PC or Mac data stations
    Data Storage
    Calender/clock functions; 64 assay protocols
    Duration
    0 - 999 s
    Filters
    415, 450, 490, 595, 655, and 750 nm
    Languages
    4 languages (English, Chinese, Japanese, Russian); LCD indication supported; printout report supported
    Linearity
    ≤1.0% from 0.0–2.0 OD; ≤2.0% from 0.0–3.0 OD
    Precision
    1.0% or 0.005 OD from 0.0–2.0 OD; 1.5% from 2.0–3.0 OD
    Printer
    Built-in, thermal
    Range
    0.0–3.5 OD
    Resolution
    0.001 OD
    Shaking Speed
    Low, mid, high
    Time
    6 sec at single wavelength, 10 sec at dual wavelengths
    Voltage
    100/240 V
    W×D×H
    34.6×37.7×16.4 cm (13.6×14.8×6.5")
    Weight
    5.5 kg (12 lbs.)
    Marketsource
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Replacement Thermal Paper, 3 Rolls
    Replacement Thermal Paper, 3 Rolls
    100447-564
    Log in for availability
    Each - $123.08

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.