Orders LinkContactUsLinkComponent
Polyester Swabs, Texwipe®
Polyester Swabs, Texwipe®
Supplier:   Texwipe
Polyester Swabs, Texwipe®
Supplier:  Texwipe
Certificates
Polyester Swabs, Texwipe®
Polyester Swabs, Texwipe®
2 Options Available Below
Product Details & Documents
Sterile polyester swabs are constructed from 100% medical-grade spun polyester.
  • Gamma irradiated
  • Packaging labeled for global use
  • Lot code and expiration date clearly marked on each package
  • Certificates of Irradiation and Compliance attached to each case
  • Pre-sterilized swabs eliminate repackaging, documentation, and validation of autoclaved swabs
These swabs are bonded securely to a polystyrene shaft by an aqueous-based adhesive. The swabs are made to exacting and consistent tolerances using high-precision automated processes. The easy-to-open, individually wrapped swabs ensure sterility at point-of-use. The swabs are visible through clear packaging which also offers visibility of seal integrity.
Documents
Product Family Options
Product Information
  • Handle color
    Handle length
    Handle width
    Head length
    Head thickness
    Head width
    Availability
    Price
  • 10127-580
    Sterile Small Polyester Swabs with Polystyrene Handle
    Sterile Small Polyester Swabs with Polystyrene Handle
    White
    134.0 mm (5.276")
    2.5 mm (0.098")
    17.0 mm (0.669")
    4.6 mm (0.181")
    4.6 mm (0.181")
    Log in for availability
    Log in for availability
    Log in for availability
    Box of 50
    Box of 50 - $72.16
    $72.16
    New Customer Promotional Price
    Log in for availability
    Case of 500
    Case of 500 - $525.36
    $525.36
    New Customer Promotional Price
    Box of 50
       $72.16
    Minimum order quantity is 10
    Case of 500
       $525.36
    Minimum order quantity is 1
    Supplier Number: STX764
  • 10127-578
    Sterile Polyester Swabs with Polystyrene Handle
    Sterile Polyester Swabs with Polystyrene Handle
    White
    134.0 mm (5.276")
    2.5 mm (0.098")
    18.0 mm (0.709")
    5.8 mm (0.228")
    5.8 mm (0.228")
    Log in for availability
    Log in for availability
    Log in for availability
    Box of 50
    Box of 50 - $43.20
    $43.20
    New Customer Promotional Price
    Log in for availability
    Case of 500
    Case of 500 - $435.76
    $435.76
    New Customer Promotional Price
    Box of 50
       $43.20
    Case of 500
       $435.76
    Supplier Number: STX763