Order Entry
United States
ContactUsLinkComponent
LYFO DISK Zygosaccharomyces bailii ATCC® MYA-4549™, Microbiologics
LYFO DISK Zygosaccharomyces bailii ATCC® MYA-4549™, Microbiologics
Catalog # 89501-786
CAS Number:  
LYFO DISK Zygosaccharomyces bailii ATCC® MYA-4549™, Microbiologics
Catalog # 89501-786
Supplier Number:  01011L
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    LYFO DISK Zygosaccharomyces bailii
  • Environmentally Preferable:
  • Storage Conditions:
    Easy storage; no dry ice required; reduces shipping costs
  • Traceability:
    Assured Quality - Traceable to reference culture
  • Cat. No.:
    89501-786
  • ATCC no.:
    ATCC® MYA-4549™*
  • Supplier no.:
    01011L

Specifications

About this item

LYFO DISK® is a lyophilized pellet containing a single strain of a QC microorganism and is the most economical reference stock culture option

LYFO DISK® is known for its ease-of-use; simply re-hydrate the pellet and inoculate. LYFO DISK® is available in a re-sealable vial containing 6 pellets of a single QC microorganism strain.

Certifications: LYFO DISK® falls under the category of In-Vitro Diagnostics Medical Devices Directives for CE Mark Conformity. This product is also listed with the FDA.
Ordering information: To receive this product, customers must have a valid ATCC End User Licensing Agreement in place with Microbiologics. Registration for this agreement is only required once, per location during the first order. Instructions for how to register can be downloaded from the website. Go to the "Related Information" box, click on "Additional Product Details", then "Instructions for End User Registration with Microbiologics".

The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC® cultures.