Order Entry
United States
ContactUsLinkComponent
Premium Two-Prong Clamp, Vinyl Coated
Premium Two-Prong Clamp, Vinyl Coated
Catalog # 470165-410
Supplier:  Wards
undefined
Premium Two-Prong Clamp, Vinyl Coated
Catalog # 470165-410
Supplier:  Wards
Supplier Number:  PRCMP05

Specifications

  • Description:
    Premium Two-Prong Clamp, Vinyl Coated
  • Cat. No.:
    470165-410
  • Supplier no.:
    PRCMP05

Specifications

About this item

Vinyl Lined Extension Lab Clamp

  • Maximum clamp opening is 11 cm
  • 14 cm connecting rod.

Clamp constructed of powder coated steel, two vinyl coated prongs attached to an aluminum rod. Completely metal construction with thread fitted actuating joints and 10 mm diameter connecting rod.