About this item
Bio-Beads™ SM-2 adsorbents are analytical-grade, neutral macroporous polymeric beads with a high surface area for adsorbing organics with MW <2,000 from aqueous solutions. Mechanism: Hydrophobic interaction chromatography (HIC).
Bio-Beads™ SM-2 nonpolar resin is useful for the adsorption of nonpolar substances or surface active agents from aqueous solutions. Common applications of Bio-Beads™ SM-2 adsorbents include.
Removal of detergents such as Triton X-100. Removal of organics such as polyaromatic hydrocarbons from water. Cleanup of drugs from plasma and urine. Cleanup of biological metabolites and pesticides. Extraction of dyes and mycotoxins from food products.
These beads can be used in aqueous solution with solvents, including alcohols, petroleum ether, diethyl ether, hexane, and solvent mixtures, without expansion or contraction of the beads.
Density: 1.02 g/ml wet
Pore size: 90 Å
Surface area: 300 m²/g