ContactUsLinkComponent
Microseal® 'F' Sealing Foil, Bio-Rad
Microseal® 'F' Sealing Foil, Bio-Rad
Catalog # 101647-202
Supplier:  Bio-Rad
Microseal® 'F' Sealing Foil, Bio-Rad
Catalog # 101647-202
Supplier:  Bio-Rad
Supplier Number:  MSF1001
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Microseal® 'F' Foil, Pack of 100
  • Cat. No.:
    101647-202
  • Supplier no.:
    MSF1001
Specifications
About this item
Microseal® 'F' Foils are aluminized foil, thin enough to pierce with a pipet tip for recovery of samples from individual wells while providing an efficient barrier against evaporation from –70 to 105°C.
These foils are compatible with automated systems such as the ABI 3700 DNA analyzer. Foils are free of DNase, RNase, and human DNA.
In addition to thermal cycling applications, the foils are suitable for cold storage.