ContactUsLinkComponent
Microseal® 'A' Sealing Film, Bio-Rad
Microseal® 'A' Sealing Film, Bio-Rad
Catalog # 101647-198
Supplier:  Bio-Rad
Microseal® 'A' Sealing Film, Bio-Rad
Catalog # 101647-198
Supplier:  Bio-Rad
Supplier Number:  MSA5001
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Microseal® 'A' Film Nonadhesive Sealing Film for PCR Plates and Tubes, Pack of 50
  • Cat. No.:
    101647-198
  • Supplier no.:
    MSA5001
Specifications
About this item
Microseal® 'A' Film is a quick and effective means to seal plates and tubes for standard PCR needs. The silicone-based film conforms tightly to most vessels, including rimless polycarbonate plates and irregular surfaces.
The compliant inner layer is designed to seal tightly during cycling and release smoothly after cycling to minimize the risk of aerosol formation and cross-contamination of samples.
This film is easily cut for use with fewer than 96 wells. Microseal® 'A' Film is not designed for optical use and should not be used in real-time PCR detection systems.
The peel-away liner protects the sealing surface from contamination prior to use.