About this item
iTaq™ Universal SYBR® Green Supermix is a 2x concentrated, ready-to-use master mix optimized for dye-based quantitative PCR (qPCR) on any real-time PCR instrument using standard or fast protocols.
- Amplify genomic DNA and difficult amplicons with high efficiency
- Consistent results across real-time instrument platforms under fast or standard protocols
- Blend of passive reference dyes for universal real-time instrument compatibility
- Antibody-mediated hot-start iTaq™ DNA polymerase for rapid activation
- Freeze/thaw up to 10 times
- Standard and fast thermal cycling protocols
iTaq™ Universal SYBR® Green Supermix contains antibody-mediated hot-start iTaq™ DNA polymerase, dNTPs, MgCl2, SYBR® Green I dye, enhancers, stabilizers, and a blend of passive reference dyes (including ROX and fluorescein).
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Application:
- Environmentally Preferable: