The Quick Start™ Bradford Protein Assay is a simple, accurate assay for determining the concentration of protein in solution, with a linear range of 1.25–25 µg/mL.
The assay kits contain ready-to-use dye reagent with either bovine serum albumin (BSA) or bovine γ-globulin standards. For either standard, choose between seven prediluted concentrations (0.125, 0.25, 0.5, 0.75, 1.0, 1.5, and 2.0 mg/ml), or making your own standard series from 2mg/ml stock solutions. The Quick Start™ Bradford Dye Reagent and Protein Standards are also available separately.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Method:Bradford
- Range:1.25–25 µg/mL
- Wavelength:595 nm
- Incubation Time:> 5 min