The MAS-100 NT® and MAS-100 NT® Ex portable microbial air samplers are the leading instruments for use in critical environments. These compact yet sophisticated devices are the preferred choice for those demanding the highest quality in microbial air monitoring.
- Compact design, easy to handle
- Improved security features and enhanced connection possibilities
- Hardware and software developed according to GAMP 4
- Fully validated system using ICR plates
- Improved communication (including RS-232 and USB ports)
- Programmable start delay of up to 60 minutes allows for personnel to be out of the sampling area when the sampling starts
- Alarm and sample log records of the last 100 events can be exported or printed at any time
- Mass flow is measured and ensures a flow of 100 LPM
- Fully meets the d50 requirements using the 300 x 0.6 mm sampling head
The MAS-100 NT® systems feature a 300-hole perforated lid for increased collection efficiency and impaction speed. Both systems utilize standard 90-100mm agar plates or can be adapted to fit 50-60mm contact plates allowing for a low consumable cost and great flexibility. Sampling at 100 LPM, these systems have the highest airflow accuracy available at ± 2.5%, compared to others that can be as high as ±10%. The integrated flow sensor allows the user to freely interchange the perforated lids without affecting the accuracy or the calibration of the unit. Sampling volumes are also easily configurable between 1 and 2,000 liters.
The MAS-100 NT® Ex shares all of the same functions of the MAS-100 NT® system but is specially designed for use in explosion proof areas. The MAS-100 NT® Ex has received ATEx Conformity and can be used in zone 2 and gas groups 11A,11B, and 11C in temperature classes T1 to T4.
- Catalog No:
- EM1.09194.0001
- EM1.09191.0001
- Flow Rate:
- 100 SLPM ±2.5% (standard litres per minute)
- 100 SLPM ±2.5% (standard litres per minute)
- Sampling volume:
- 1 to 2,000 litres, in increments of 1 liter
- 1 to 2,000 litres, in increments of 1 liter
- Running Time:
- Total running time up to 7 hours
- Total running time up to 7 hours
- Display:
- Backlit liquid crystal display
- Backlit liquid crystal display
- Power:
- 110-240 V, 50-60 KHz
- 110-240 V, 50-60 KHz
- Battery Type:
- Li-ion, rechargable battery pack
- Li-ion, rechargable battery pack
- Charging Time:
- Full recharge time approx. 3.5 hours
- Full recharge time approx. 3.5 hours
- Dimensions:
- Height 25 cm
Diameter 11 cm - Height 25 cm
Diameter 11 cm
- Weight:
- 2.38 kg
- 2.38 kg