About this item
The Thermo Scientific Varioskan LUX combines simplicity with top-end performance
Ideal for applications including absorbance (UV-Vis), fluorescence intensity, luminescence, AlphaScreen, AlphaLISA, and time-resolved fluoresence.
Plate types for various applications include:
Absorbance: 6-384 well plates
Fluorescence: 6-1536 well plates
Time-Resolved Fluorescence: 6-1536 well plates
Luminescence: 6-1536 well plates (spectral scanning 6-384 well plates)
AlphaScreen: 6-1536 well plates
SkanIt Software included. A microplate reader with so many automated features requires a truly user-friendly interface. Enter the newly designed 4th generation SkanIt software. Its easy-to-navigate interface will guide you through the measurement process and getting the results you need. SkanIt is available in two editions: Research Edition for scientists working in life science research, and Drug Discovery Edition that provides features to help you comply with the requirements of FDA 21 CFR Part 11. SkanIt software is available in nine languages: English, French, German, Spanish, Portuguese, Italian, Chinese (simplified), Japanese and Russian.
Designed for researchers with a wide variety of needs, Varioskan LUX offers features to save time and help reduce common errors in the lab - including automated dynamic range selection, which adjusts the optimal reading range based on signal intensities, and built-in safety controls, which alert you to potential errors before they begin