Specifications
- Antibody Type:Primary
- Antigen Name:Nerve Growth Factor beta
- Antigen Symbol:NGFB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Birds,Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µL
- Epitope:Multiple points of reactivity likely because of the polyclonal nature of the antibody
- Cross Adsorption:No
- Neutralisation:Yes
- Format:Lyophilized
- Antigen Synonyms:Beta-nerve growth factor
- Immunogen:Native mouse beta NGF purified from submaxillary salivary gland (95% purity by PAGE)
- Cat. No.:10782-710
- Supplier no.:R-085-100
Specifications
About this item
FUNCTION: Nerve growth factor is important for the development and maintenance of the sympathetic and sensory nervous systems. It stimulates division and differentiation of sympathetic and embryonic sensory neurons. SUBUNIT: Homodimer, associated by noncovalent forces. SUBCELLULAR LOCATION: Secreted protein. SIMILARITY: Belongs to the NGF-beta family.
A cross reactivity of less than 1% to recombinant human BDNF, NT3, NT4/5 by ELISA has been shown.
Application Information:
IHC, 1-site ELISA, WB, immunoblot, inhibition of biological activity. A dilution of 1:1000-1:5000 is recommended for IHC, western blot and immunoblot; 1:15000 for ELISA; for inhibition of biological activity: 1:10-50 for in vitro, 5-10 µl/g body weight for in vivo. This antiserum completely inhibits neuronal survival and the outgrowth actions of murine NGF in chicken DRG in vitro. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: beta NGF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple points of reactivity likely because of the polyclonal nature of the antibody
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat, Human, Avis