Specifications
- Antibody Type:Primary
- Antigen Name:Presenilin 1 loop region
- Antigen Symbol:PSNL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western Blot:Yes
- Size:500 µg
- Epitope:Human PS1 [301-317] in the loop region
- Cross Adsorption:No
- Format:Lyophilized
- Antigen Synonyms:Presenilin 1|PSNL1
- Immunogen:A synthetic peptide (GDPEAQRRVSKNSKYNA-C) corresponding to human PS1 [301-317] in the loop region conjugated via additional C-terminal Cys to Diphtheria toxoid.
- Cat. No.:10782-552
- Supplier no.:R-1680-500
Specifications
About this item
Presenilin-1 (PSEN1) is a multi-pass membrane protein and component of the gamma-secretase complex. PSEN1 is thought to play a role in intracellular signaling and gene expression or in linking chromatin to the nuclear membrane. It may also play a role in hematopoiesis. Defects in PSEN1 are a cause of Alzheimer disease type 3 (AD3), a familial early-onset form of Alzheimer disease (Ref:SWISS-Prot).
Confirmed by Western blotting using transfected cells, presenilin 1 knock-out mouse cells and mouse and human brain.
Application Information:
Western Blot. Suggested dilution of 1:1,000 is recommended for WB. Full length presenilin 1 (467 aa) has relative MW of about 45 kDa, with this antibody most commonly detected as cleaved CTF of 19 kDa. Human or mouse brain samples commonly prepared with reducing agent (50mM DTT), urea (2.3 M), SDS (1.5%) in 62.5 mM Tris-HCL pH 6.8 sample buffer (without boiling) heating to 50 C for 15 min. Biosensis recommends that the optimal working dilution should be determined by the end user.
Type: Primary
Antigen: Presenilin 1 loop region
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Human PS1 [301-317] in the loop region
Host: Rabbit
Isotype: IgG
Reactivity: