- Antibody Type:Primary
- Antigen Name:Endomorphin-1 And Endomorphin-2
- Antigen Symbol:EM1 + EM2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:50 µg
- Cross Adsorption:No
- Format:Lyophilized
- Antigen Synonyms:EM 2|EM 1
- Immunogen:Antibodies to human Endomorphins 1 and 2 were made by immunising rabbits with the human Endomorphin-1 peptide (YPWF) and then selecting those antibodies that cross-reacted with both human Endomorphins 1 and 2.
- Cat. No.:10782-492
- Supplier no.:R-1495-50
Endomorphins 1 and 2 are endogenous opioid peptides which have the highest affinity for the mu-opioid receptors. Located in various parts of the brain and interacts with mu-opioid receptors and produces analgesia.
Antibody recognizes Endomorphins. A dilution of 5-10 µg/ml is recommended for immunohistochemistry using formalin fixed and paraffin embedded tissues and for 4% paraformaldehyde fixed frozen tissues. A dilution of 5-15 µg/ml is recommended for immunofluorescence. . Biosensis recommends optimal dilutions/concentrations should be determined by the end user. Cross-reactivity Human; mouse; rat. Endomorphin-1 is highly conserved molecule so cross-reactivity with other species is expected. Cross-reactivity with other opioid peptides is as follows: with Endomorphin-2 = 70%; Met-enkephalin <0.03%; with Leu-enkephalin 0.01%; with beta-endorphin<0.1%.
Application Information:
A dilution of 5-10 µg/ml is recommended for immunohistochemistry using formalin fixed and paraffin embedded tissues and for 4% paraformaldehyde fixed frozen tissues. A dilution of 5-15 µg/ml is recommended for immunofluorescence. . Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: Endomorphin-1 and Endomorphin-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat