Anti-Protocadherin alpha C2 Mouse Monoclonal Antibody [clone: 3D12]
Catalog # 10782-272
Supplier: Avantor
Specifications
- Antibody Type:Primary
- Antigen Name:Protocadherin Alpha C2
- Antigen Symbol:PCDHAC2
- Clonality:Monoclonal
- Clone:3D12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Format:Lyophilized
- Antigen Synonyms:PCDHAC2|PCDH-alpha-C2
- Immunogen:Partial recombinant human Protocadherin alpha C2 [81-190) with a GST tag.
- Cat. No.:10782-272
- Supplier no.:M-904-100
Specifications
About this item
Protocadherin alpha-C2 is 1 of 15 tandemly arranged genes within the PCDHA gene cluster. These 15 genes function as 'variable' exons that are individually spliced to a downstream constant region to form distinct PCDHA transcripts. They encode cadherin-like cell surface proteins that are expressed in neurons and are present at synaptic junctions.
Specificity has been confirmed by WB and ELISA against the antigen.
Application Information:
This antibody is recommended for WB and ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: Protocadherin alpha C2
Clonality: Monoclonal
Clone: 3D12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human