Orders LinkContactUsLinkComponent
Power Function Generator
Power Function Generator
Power Function Generator
Catalog # 470162-794
Supplier #:  1009956
Power Function Generator
Catalog # 470162-794
Power Function Generator
Power Function Generator
Power Function Generator
Price
$1,476.80
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Compact function generator
  • With retractable feet.
  • Includes power supply
  • Input voltage 110 V
Featuring illuminated, digital display for frequency, signal form, offset and other parameters. The output is short-circuit protected as well as being protected against induced voltages and spark discharges, for when experiment leads are unintentionally pulled out while coils are connected. In internal sweep mode, one trigger pulse is output per cycle and the voltage output is proportional to the frequency.
Specifications
  • Description:
    Power Function Generator