Orders LinkContactUsLinkComponent
Laser Reflection Sensor
Laser Reflection Sensor
Laser Reflection Sensor
Catalog # 470162-778
Supplier #:  1001034
Laser Reflection Sensor
Catalog # 470162-778
Laser Reflection Sensor
Laser Reflection Sensor
Laser Reflection Sensor
Price
$374.69
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Sensor for triggering the digital counter in time measurements on moving objects. Suitable for opto electronic scanning of light and dark markings on moving objects or in conjunction with a reflecting foil to form a wide spaced obstruction sensor. The intensity of the laser beam is adjusted automatically according to the distance of the object. Includes reflecting foil, stand rod with screw thread, and connector lead with 8 pin miniDIN plugs.
Maximum range: 2.5 m
Laser power: <1 mW
Laser protection class: II
Specifications
  • Description:
    Laser Reflection Sensor