ContactUsLinkComponent
The Chemistry of Invisible Inks STEM Kit
The Chemistry of Invisible Inks STEM Kit
  470213-360
undefined
The Chemistry of Invisible Inks STEM Kit
  470213-360
 :  AISINKIT
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    The Chemistry of Invisible Inks STEM Kit
  • No. of students:
    40
  • Cat. No.:
    470213-360
  • Includes:
    DVD with PDF Teacher and Student Guides, Digital Content
  • Supplier no.:
    AISINKIT
Explore How Invisible Inks Are Made
  • Inquiry-based hands-on laboratory
  • Guided and independent investigations
  • STEM Integration
This kit is a unique teaching tool for introducing your students to the fascinating chemistry and physics behind invisible inks, i.e. colorless substances usable for writing that can be made to turn to color using a chemical or physical process. Students investigate how organic fluids and chemical reagents can be used as invisible inks. Relying on historical example, students create various invisible inks (and security) documents for other student groups to analyze and decode! Includes materials for two detailed activities (each with a ‘model’ and ‘inquiry’ investigation paths – a total of four investigational activities). Covered concepts include: data analysis, chemical & physical processes, elements, mixtures & compounds, pH indicators, transmitted vs. reflected light, the periodic table, fluorescent compounds, types of chemical reactions, and hypothesis testing.
The kit includes materials for two detailed activities (each with a ‘model’ and ‘inquiry’ investigation paths for a total of four investigational activities). Covered concepts include data analysis, chemical and physical processes, elements, mixtures and compounds, pH indicators, transmitted versus reflected light, the periodic table, fluorescent compounds, types of chemical reactions, and hypothesis testing.