ContactUsLinkComponent
pH Indicators and Dyes STEM Kit
pH Indicators and Dyes STEM Kit
Catalog # 470213-356
undefined
pH Indicators and Dyes STEM Kit
Catalog # 470213-356
Supplier Number:  AISPHKIT
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    pH Indicators and Dyes STEM Kit
  • Environmentally Preferable:
  • No. of lab groups:
    20
  • Cat. No.:
    470213-356
  • Includes:
    Instructional CD with PDF Student and Teachers Guide
  • Supplier no.:
    AISPHKIT
Specifications
About this item
This kit is a unique teaching tool for introducing students to the chemistry behind plant pigments (the blueberry bio-pigment cyanidin) and how they can be used as pH indicators and fabric dyes.
  • Kit Contains All Required Chemicals and Supplies
  • Students Design Test Strips and Compare to Those Commercially Available
  • Material Accommodates up to Forty Students
  • Covered Concepts Include Data Analysis, Accuracy vs. Precision, Bio-Pigment Structure, and More
  • Teacher's Guide Included
They use their knowledge to create their own pH test strips and evaluate them against a commercial type.
Covered concepts include data analysis, accuracy vs. precision, bio-pigment structure, pH, pH scale, pH indicators, stains and dyes, acids, bases, acid-base interactions, solutions, and self-ionization of water.
Ordering information: This kit includes four detailed activities featuring 'model' and 'inquiry' investigation paths for 11 investigational activities, which can accommodate up to 40 students. It also contains a DVD with PDFs of teacher and student guides and other digital content.