Growth regulated protein beta (GRO-beta), also known as CXCL2, is a chemokine that is secreted by macrophages, neutrophils, and monocytes at sites of inflammation. GRO-beta functions as a chemoattractant for leukocytes and hematopoietic stem cells. GRO-beta activity is mediated through binding the G-protein-coupled chemokine receptor CXCR2.
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions. This product is for research use only and is not for use in humans.
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Biological Activity:Bioactivity: No biological activity data is available at this time
- Gene ID:P19875
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:N
- Protein Synonyms:CXCL2|GRO2|MIP-2α|MGSAβ
- Protein/Peptide Name:GRO-beta
- Purity:≥95%
- Molecular Weight:7.9 kDa
- Sequence:APLATELRCQ CLQTLQGIHL KNIQSVKVKS PGPHCAQTEV IATLKNGQKA CLNPASPMVK KIIEKMLKNG KSN
- Endotoxin Level:Low
- Formulation:0.1% Trifluoroacetic Acid (TFA)
- Nuclease-free:N
- Shipping Temperature:RT
- Grade:RUO