Anti-SLC5A5 Mouse Monoclonal Antibody [clone: FP5]
10737-774
: Abnova
- Antibody Type:Primary
- Antigen Name:Solute Carrier Family 5 (low affinity glucose cotransporter) Member 5
- Antigen Symbol:SLC5A5
- Clonality:Monoclonal
- Clone:FP5
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Format:Liquid
- Gene ID:6528
- Antigen Synonyms:TDH1|NIS|Sodium-Iodide Symporter|CNGA1/3
- Amino Acid Number:468 to 643
- Storage Buffer:PBS, pH 7.4 (50% glycerol)
- Storage Temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant mannose fusion protien corresponding to amino acids 468-643 of human SLC5A5.
- Purification:Protein G purification
- Cat. No.:10737-774
- Supplier no.:MAB11494
Mouse monoclonal antibody raised against recombinant of SLC5A5.
Recommended Dilutions: Immunofluorescence Immunohistochemistry Western Blot: 1:1000
Type: Primary
Antigen: SLC5A5
Clonality: Monoclonal
Clone: FP5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: