Anti-CBX beta Chicken Polyclonal Antibody
Catalog # 10745-294
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Symbol:Clostridium botulinum Type B Neurotoxin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Chicken
- Isotype:IgY
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Format:Liquid
- Storage Buffer:PBS, pH 7.2 (0.1% sodium azide).
- Storage Temperature:Store at 4°C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified Clostridium botulinum Type B Neurotoxin emulsified in Freund's adjuvant.
- Purification:Affinity purification
- Cat. No.:10745-294
- Supplier no.:PAB29146
Specifications
About this item
Chicken polyclonal antibody raised against Clostridium botulinum Type B Neurotoxin.
Recommended Dilutions: Western Blot: 1:1000-1:200
Type: Primary
Antigen: CBX beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: