Order Entry
United States
ContactUsLinkComponent
Anti-PPP1R13B Mouse Monoclonal Antibody [clone: 6H1]
Anti-PPP1R13B Mouse Monoclonal Antibody [clone: 6H1]
Catalog # 10597-866
Supplier:  Abnova
Anti-PPP1R13B Mouse Monoclonal Antibody [clone: 6H1]
Catalog # 10597-866
Supplier:  Abnova
Supplier Number:  H00023368-M04

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Protein Phosphatase 1, Regulatory Subunit 13B
  • Antigen Symbol:
    PPP1R13B
  • Clonality:
    Monoclonal
  • Clone:
    6H1
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG2a kappa
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Cross Adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    23368
  • Antigen Synonyms:
    p53BP2-like|ASPP1|p85
  • Amino Acid Number:
    1 to 90
  • Storage Buffer:
    1x PBS, pH 7.4
  • Sequence:
    MMPMILTVFLSNNEQILTEVPITPETTCRDVVEFCKEPGEGSCHLAEVWRGNERPIPFDHMMYEHLQKWGPRREEVKFFLRHEDSPTENS
  • Storage Temperature:
    Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    PPP1R13B (NP_056131.2, 1 a.a. ~ 90 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
  • Cat. No.:
    10597-866
  • Supplier no.:
    H00023368-M04

Specifications

About this item

Mouse monoclonal antibody raised against a partial recombinant PPP1R13B.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: PPP1R13B
Clonality: Monoclonal
Clone: 6H1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: 0