Anti-DUX4 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: P2B1]
Catalog # 10711-416
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Double Homeobox 4
- Antigen Symbol:DUX4
- Clonality:Monoclonal
- Clone:P2B1
- Conjugation:RPE (R-Phycoerythrin)
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Format:Liquid
- Gene ID:22947
- Antigen Synonyms:double homeobox protein 10|DUX10|double homeobox protein 4/10|double homeobox protein DUX10|double homeobox protein 4
- Storage Buffer:PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
- Storage Temperature:Store at 4°C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant GST fusion protein corresponding to a 76 amino acid fragment at C-terminus of human DUX4.
- Purification:Protein G purification
- Cat. No.:10711-416
- Supplier no.:MAB11458
Specifications
About this item
Mouse monoclonal antibody raised against recombinant partial DUX4.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: DUX4
Clonality: Monoclonal
Clone: P2B1
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human