Anti-NKX2-8 Rabbit Polyclonal Antibody
10727-232
: Abnova
- Antibody Type:Primary
- Antigen Name:NK2 homeobox 8
- Antigen Symbol:NKX2-8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Format:Liquid
- Gene ID:26257
- Antigen Synonyms:NKX2-8|Homeobox protein NK-2 homolog H|NKX2.8|NKX-2.8|NKX2G|Nkx2-9|NKX2H
- Storage Buffer:PBS (0.02% sodium azide)
- Storage Temperature:Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to 19 amino acids at internal region of human NKX2-8.
- Purification:Peptide affinity purification
- Cat. No.:10727-232
- Supplier no.:PAB25801
Rabbit polyclonal antibody raised against synthetic peptide of NKX2-8.
Recommended Dilutions: Western Blot: 1 ?g/ml; Immunohistochemistry: 2.5 ?g/ml
Type: Primary
Antigen: NKX2-8
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human, Mouse, Rat