Anti-WDR5 Rabbit Polyclonal Antibody
Catalog # 10744-510
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:WD Repeat Domain 5
- Antigen Symbol:WDR5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Format:Liquid
- Gene ID:11091
- Antigen Synonyms:BMP2-induced 3-kb gene protein|BIG3|WDR5|SWD3|WD repeat-containing protein 5|BIG-3
- Storage Buffer:PBS (0.02% sodium azide)
- Storage Temperature:Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to N-terminus of human WDR5.
- Purification:Affinity purification
- Cat. No.:10744-510
- Supplier no.:PAB25494
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of WDR5.
Recommended Dilutions: Western Blot: 1 ?g/ml; Immunocytochemistry: 5 ?g/ml
Type: Primary
Antigen: WDR5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human