Anti-RELA Rabbit Polyclonal Antibody
10675-280
: Abnova
- Antibody Type:Primary
- Antigen Name:RELA proto-oncogene, NF-kB subunit
- Antigen Symbol:RELA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Format:Liquid
- Gene ID:5970
- Antigen Synonyms:nuclear factor NF-kappa-B p65 subunit|v-rel reticuloendotheliosis viral oncogene homolog A|p65|nuclear factor of kappa light polypeptide gene enhancer in B-cells 3|transcription factor p65|v-rel avian reticuloendotheliosis viral oncogene homolog A (nuclear factor of kappa light polypeptide gene enhancer in B-cells 3 (p65))
- Modification:phospho-Ser536
- Storage Buffer:PBS (without Mg(2+) and Ca(2+)), 150 mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide)
- Sequence:F-S-Sp-I-A
- Storage Temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Synthetic phosphopeptide corresponding to residues surrounding S536 of human RELA.
- Purification:purified by Affinity Chromatography
- Cat. No.:10675-280
- Supplier no.:PAB25235
Rabbit polyclonal antibody raised against synthetic phosphopeptide of RELA.
Recommended Dilutions: Immunohistochemistry: 1:50-1:100; Western Blot: 1:500-1:1000; Immunofluorescence: 1:100-1:200
Type: Primary
Antigen: RELA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat