Anti-HFE Rabbit Polyclonal Antibody
Catalog # 10717-322
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Hemochromatosis
- Antigen Symbol:HFE
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:100 µL
- Cross Adsorption:No
- Format:Liquid
- Gene ID:3077
- Antigen Synonyms:TFQTL2|hereditary hemochromatosis protein HLA-H|hereditary hemochromatosis protein|MVCD7|MHC class I-like protein HFE|HLA-H|HFE1|high Fe|HH
- Storage Buffer:PBS, pH 7.2 (0.05% sodium azide)
- Storage Temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to HFE.
- Purification:Affinity purification
- Cat. No.:10717-322
- Supplier no.:PAB25214
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of HFE.
Recommended Dilutions: Western Blot: 1:500-1:1000; Immunohistochemistry: 1:50-1:200
Type: Primary
Antigen: HFE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human