Anti-MAP2K6 Mouse Monoclonal Antibody [clone: 3H12C8]
Catalog # 10724-706
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Mitogen-Activated Protein Kinase Kinase 6
- Antigen Symbol:MAP2K6
- Clonality:Monoclonal
- Clone:3H12C8
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µL
- Cross Adsorption:No
- Format:Liquid
- Form:Ascites
- Gene ID:5608
- Antigen Synonyms:PRKMK6|MKK6|MAPKK6|SAPKK-3|MEK6|SAPKK3
- Storage Buffer:0.03% sodium azide
- Storage Temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to human MAP2K6.
- Cat. No.:10724-706
- Supplier no.:MAB10794
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant MAP2K6.
Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunofluorescence: 1:200-1:1000; Flow Cytometry: 1:200-1:400
Type: Primary
Antigen: MAP2K6
Clonality: Monoclonal
Clone: 3H12C8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse