Anti-KLHL11 Mouse Monoclonal Antibody [clone: 1B9A8]
Catalog # 10722-668
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Kelch-like 11
- Antigen Symbol:KLHL11
- Clonality:Monoclonal
- Clone:1B9A8
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 µL
- Cross Adsorption:No
- Format:Liquid
- Form:Ascites
- Gene ID:55175
- Storage Buffer:0.03% sodium azide
- Storage Temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to human KLHL11.
- Cat. No.:10722-668
- Supplier no.:MAB10717
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant KLHL11.
Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunofluorescence: 1:200-1:1000; Flow Cytometry: 1:200-1:400
Type: Primary
Antigen: KLHL11
Clonality: Monoclonal
Clone: 1B9A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human