Order Entry
United States
ContactUsLinkComponent
 
Anti-FGG Mouse Monoclonal Antibody [clone: 5A6]
  10714-714
 :  Abnova
undefined
Anti-FGG Mouse Monoclonal Antibody [clone: 5A6]
  10714-714
 :  Abnova
 :  MAB10649
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    fibrinogen gamma chain
  • Antigen Symbol:
    FGG
  • Clonality:
    Monoclonal
  • Clone:
    5A6
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 µL
  • Environmentally Preferable:
  • Cross Adsorption:
    No
  • Format:
    Liquid
  • Form:
    Ascites
  • Gene ID:
    2266
  • Storage Buffer:
    0.03% sodium azide
  • Storage Temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to human FGG.
  • Cat. No.:
    10714-714
  • Supplier no.:
    MAB10649

 

 

Mouse monoclonal antibody raised against partial recombinant FGG.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunofluorescence: 1:200-1:1000

Type: Primary
Antigen: FGG
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: 5A6
Epitope:
Host: IgG2b
Isotype: Mouse
Reactivity: Human