Anti-AKAP13 Rabbit Polyclonal Antibody
10692-368
: Abnova
- Antibody Type:Primary
- Antigen Name:A-kinase anchor protein 13
- Antigen Symbol:AKAP13
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:100 µL
- Cross Adsorption:No
- Format:Liquid
- Gene ID:11214
- Antigen Synonyms:Breast cancer nuclear receptor-binding auxiliary protein|Lymphoid blast crisis oncogene|p47|A-kinase anchor protein 13|AKAP-Lbc|LBC|Non-oncogenic Rho GTPase-specific GTP exchange factor|Protein kinase A-anchoring protein 13|LBC oncogene|AKAP-13|Guanine nucleotide exchange factor Lbc|BRX|AKAP13|PRKA13|HT31|Human thyroid-anchoring protein 31
- Storage Buffer:PBS, pH 7.2 (0.05% sodium azide)
- Storage Temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to AKAP13.
- Purification:Antigen affinity purification
- Cat. No.:10692-368
- Supplier no.:PAB24967
Rabbit polyclonal antibody raised against synthetic peptide of AKAP13.
Recommended Dilutions: Western Blot: 1:500-1:1000; Immunohistochemistry: 1:50-1:200
Type: Primary
Antigen: AKAP13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human