Anti-RCAN1 Goat Polyclonal Antibody
10734-030
: Abnova
- Antibody Type:Primary
- Antigen Name:Regulator of Calcineurin 1
- Antigen Symbol:RCAN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Mouse
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Format:Liquid
- Gene ID:1827
- Antigen Synonyms:ADAPT78|DSCR1|Regulator of calcineurin 1|RCAN1|DSC1|Calcipressin-1|Myocyte-enriched calcineurin-interacting protein 1|Adapt78|CSP1|Down syndrome critical region protein 1
- Storage Buffer:Tris saline, pH 7.3 (0.5 % BSA, 0.02% sodium azide)
- Sequence:CHIGSSHLAPPNPD
- Storage Temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to internal region of human RCAN1.
- Purification:Antigen affinity purification
- Cat. No.:10734-030
- Supplier no.:PAB24686
Goat polyclonal antibody raised against synthetic peptide of RCAN1.
Recommended Dilutions: ELISA: 1:32000; Western Blot: 1-3 ?g/ml
Type: Primary
Antigen: RCAN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse