Anti-ERBB2 Rabbit Polyclonal Antibody
Catalog # 10674-594
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Erb-b2 receptor tyrosine kinase 2
- Antigen Symbol:ERBB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Format:Liquid
- Gene ID:2064
- Antigen Synonyms:NEU|TKR1|MLN 19|CD340|HER2|HER-2/neu|HER-2|NGL
- Modification:phospho-Tyr877
- Storage Buffer:PBS (without Mg(2+) and Ca(2+)), 150 mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide)
- Sequence:T-E-Yp-H-A
- Storage Temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Synthetic phosphopeptide corresponding to residues surrounding Y877 of human ERBB2.
- Purification:purified by Affinity Chromatography
- Cat. No.:10674-594
- Supplier no.:PAB25294
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic phosphopeptide of ERBB2.
Recommended Dilutions: Immunohistochemistry: 1:50-1:100; Western Blot: 1:500-1:1000; Immunofluorescence: 1:100-1:200
Type: Primary
Antigen: ERBB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat