Anti-RAB39B Mouse Monoclonal Antibody [clone: 3B7]
Catalog # 10598-894
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Ras-related protein Rab-39B
- Antigen Symbol:RAB39B
- Clonality:Monoclonal
- Clone:3B7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Format:Liquid
- Gene ID:116442
- Antigen Synonyms:MRX72|WSMN
- Amino Acid Number:1 to 213
- Storage Buffer:1x PBS, pH 7.4
- Sequence:MEAIWLYQFRLIVIGDSTVGKSCLIRRFTEGRFAQVSDPTVGVDFFSRLVEIEPGKRIKLQIWDTAGQERFRSITRAYYRNSVGGLLLFDITNRRSFQNVHEWLEETKVHVQPYQIVFVLVGHKCDLDTQRQVTRHEAEKLAAAYGMKYIETSARDAINVEKAFTDLTRDIYELVKRGEITIQEGWEGVKSGFVPNVVHSSEEVVKSERRCLC
- Storage Temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:RAB39B (AAH09714, 1 a.a. ~ 213 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
- Cat. No.:10598-894
- Supplier no.:H00116442-M02
Specifications
About this item
Mouse monoclonal antibody raised against a full-length recombinant RAB39B.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: RAB39B
Clonality: Monoclonal
Clone: 3B7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human