Anti-FANCF Mouse Polyclonal Antibody
10556-344
: Abnova
- Antibody Type:Primary
- Antigen Name:Fanconi Anemia Complementation Group F
- Antigen Symbol:FANCF
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Cross Adsorption:No
- Format:Liquid
- Gene ID:2188
- Antigen Synonyms:FAF
- Amino Acid Number:1 to 374
- Storage Buffer:1x PBS, pH 7.4
- Sequence:MESLLQHLDRFSELLAVSSTTYVSTWDPATVRRALQWARYLRHIHRRFGRHGPIRTALERRLHNQWRQEGGFGRGPVPGLANFQALGHCDVLLSLRLLENRALGDAARYHLVQQLFPGPGVRDADEETLQESLARLARRRSAVHMLRFNGYRENPNLQEDSLMKTQAELLLERLQEVGKAEAERPARFLSSLWERLPQNNFLKVIAVALLQPPLSRRPQEELEPGIHKSPGEGSQVLVHWLLGNSEVFAAFCRALPAGLLTLVTSRHPALSPVYLGLLTDWGQRLHYDLQKGIWVGTESQDVPWEELHNRFQSLCQAPPPLKDKVLTALETCKAQDGDFEVPGLSIWTDLLLALRSGAFRKRQVLGLSAGLSSV
- Storage Temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:FANCF (NP_073562.1, 1 a.a. ~ 374 a.a) full-length human protein.
- Cat. No.:10556-344
- Supplier no.:H00002188-B01P
Mouse polyclonal antibody raised against a full-length human FANCF protein.
Recommended Dilutions: Western Blot: 1 ?g/ml
Type: Primary
Antigen: FANCF
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Mouse
Reactivity: Human